Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A109139-100MG
A109139-250MG
A109139-1G
A109139-5G
76796-724EA
6
USD
76796-724
76796-726
77666-588
77878-825
Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate 97%
Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₉N₃O₂ |
MDL Number:
MFCD02186164 CAS Number: 64951-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...