Manidipine dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[4-(Diphenylmethyl)-1-piperazinyl]ethyl Methyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarb, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-[4-(Diphenylmethyl)-1-piperazinyl]ethyl Methyl Ester Dihydrochloride, 3-(2-(4-Benzhydrylpiperazin-1-yl)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate dihydrochloride
A108830-100MG
A108830-10MG
A108830-1G
A108830-250MG
A108830-50MG
A108830-5G
76796-676EA
12
USD
76796-676
76796-678
76796-680
76796-682
76796-684
76796-686
Manidipine dihydrochloride 98%
Manidipine dihydrochloride
Precaución: for Research Use Only
Formula:
C₃₅H₃₈N₄O₆ MW: 683.63 g/mol |
MDL Number:
MFCD00896434 CAS Number: 89226-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...