Ethyl 2-(5-(4-methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)acetate 97%

Supplier: Ambeed
Warning

A108739-100MG A108739-1G A108739-250MG A108739-5G
76796-668EA 16 USD
76796-668 76796-670 76796-672 76796-674
Ethyl 2-(5-(4-methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)acetate 97%
Ethyl 2-(5-(4-methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)acetate

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₄O₂
MDL Number: MFCD10565679
CAS Number: 402948-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR