Methyl indole-7-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-indole-7-carboxylate

A108524-100G A108524-1G A108524-5G
76796-654EA 446.64 USD
76796-654 76796-656 76796-658
Methyl indole-7-carboxylate 97%
Methyl indole-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00211064
CAS Number: 93247-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR