3-Fluoro-4-cyanophenyl-4-ethylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-3-fluorophenyl 4-Ethylbenzoate, 4-Ethylbenzoic acid 4-Cyano-3-fluorophenyl ester

A107794-10G A107794-5G A107794-25G
76796-628EA 28 USD
76796-628 76796-630 76796-632 77878-816
3-Fluoro-4-cyanophenyl-4-ethylbenzoate 97%
3-Fluoro-4-cyanophenyl-4-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₂FNO₂
MDL Number: MFCD00689047
CAS Number: 86776-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR