Laurocapram 97%

Supplier: Ambeed
Warning

Synonyms: N-Dodecyl-ε-caprolactam, Azone, Laurocapram, N-Dodecyl-epsilon-caprolactam, 1-Dodecylazepan-2-one

A107258-100G A107258-25G A107258-500G A107258-1000G
76796-610EA 13 USD
76796-610 77549-062 77666-562 77878-813
Laurocapram 97%
Laurocapram

Precaución: for Research Use Only

Formula: C₁₈H₃₅NO
MW: 281.48 g/mol
MDL Number: MFCD00190580
CAS Number: 59227-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR