DIPSO sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 3-(bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonate

A107091-100G A107091-25G A107091-5G
76796-580EA 105.03 USD
76796-580 76796-582 76796-584
DIPSO sodium salt 98%
DIPSO sodium salt

Precaución: for Research Use Only

Formula: C₇H₁₆NNaO₆S
MW: 265.2598 g/mol
MDL Number: MFCD00070010
CAS Number: 102783-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR