Diethyl (4-bromobenzyl)phosphonate 95%

Supplier: Ambeed
Warning

Synonyms: Diethyl 4-bromobenzylphosphonate, (4-Bromobenzyl)phosphonic acid diethyl ester

A106906-100G A106906-1G A106906-5G A106906-25G A106906-10G
76796-574EA 191 USD
76796-574 76796-576 76796-578 77565-874 77570-422
Diethyl (4-bromobenzyl)phosphonate 95%
Diethyl (4-bromobenzyl)phosphonate

Precaución: For research use only.

Formula: C₁₁H₁₆BrO₃P
MDL Number: MFCD05663693
CAS Number: 38186-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR