Diphenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Diphenylacetic acid

A106893-1000G A106893-10G A106893-25G A106893-500G A106893-100G
76796-566EA 186.72 USD
76796-566 76796-568 76796-570 76796-572 77549-052
Diphenylacetic acid 98%
Diphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00004251
CAS Number: 117-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR