Dimethoxy(diisopropyl)silane 95+%

Supplier: Ambeed
Danger

Synonyms: Diisopropyldimethoxysilane

A106340-25G A106340-5G
76796-546EA 23.34 USD
76796-546 76796-548
Dimethoxy(diisopropyl)silane 95+%
Dimethoxy(diisopropyl)silane

Precaución: for Research Use Only

Formula: C₈H₂₀O₂Si
MW: 176.33 g/mol
MDL Number: MFCD00239343
CAS Number: 18230-61-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR