H-D-Thr(tBu)-OMe·HCl 97%

Supplier: Ambeed
Warning

A106071-1G A106071-250MG
76796-530EA 17 USD
76796-530 76796-532
H-D-Thr(tBu)-OMe·HCl 97%
H-D-Thr(tBu)-OMe·HCl

Precaución: for Research Use Only

Formula: C₉H₂₀ClNO₃
MDL Number: MFCD00237278
CAS Number: 115141-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR