(2R,3S)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate 90%

Supplier: Ambeed
Warning

A105847-100G A105847-250MG
76796-512EA 479 USD
76796-512 76796-514
(2R,3S)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate 90%
(2R,3S)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate

Precaución: for Research Use Only

Formula: C₂₁H₂₁ClO₅
MDL Number: MFCD00079148
CAS Number: 3601-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR