Hydroxychloroquine sulfate 99%

Supplier: Ambeed
Warning

A105779-100G A105779-10G A105779-1G A105779-250MG A105779-25G A105779-5G A105779-50MG
76796-498EA 347 USD
76796-498 76796-500 76796-502 76796-504 76796-506 76796-508 76796-510 77666-550
Hydroxychloroquine sulfate 99%
Hydroxychloroquine sulfate

Precaución: for Research Use Only

Formula: C₁₈H₂₆ClN₃O
MDL Number: MFCD00078203
CAS Number: 747-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR