N-(tert-Butoxycarbonyl)-1,5-diaminopentane 98%

Supplier: Ambeed
Danger

Synonyms: N-Boc-1,5-diaminopentane, tert-Butyl N-(5-Aminopentyl)carbamate, tert-Butyl (5-aminopentyl)carbamate, N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, NH2-C5-NH-Boc, N-Boc-1,5-pentanediamine

A105669-1G A105669-25G A105669-10G A105669-5G
76796-494EA 25.47 USD
76796-494 76796-496 77549-012 77549-014
N-(tert-Butoxycarbonyl)-1,5-diaminopentane 98%
N-(tert-Butoxycarbonyl)-1,5-diaminopentane

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂O₂
MW: 202.30 g/mol
MDL Number: MFCD00210020
CAS Number: 51644-96-3
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR