Galaxolide(1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran) ∼ 50% in diethyl phthalate

Supplier: Ambeed
Danger

A105200-25G A105200-100G
76796-484EA 14 USD
76796-484 76796-486 77878-808
Galaxolide(1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran) ∼ 50% in diethyl phthalate
Galaxolide(1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran)

Precaución: for Research Use Only

Formula: C₁₈H₂₆O
MDL Number: MFCD00217003
CAS Number: 1222-05-5
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR