4-(Pyrrol-1-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-(1H-Pyrrol-1-yl)benzaldehyde

A105138-10G A105138-1G A105138-250MG A105138-25G A105138-5G
76796-474EA 147 USD
76796-474 76796-476 76796-478 76796-480 76796-482
4-(Pyrrol-1-yl)benzaldehyde 97%
4-(Pyrrol-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MW: 171.195 g/mol
MDL Number: MFCD03822126
CAS Number: 23351-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR