Methyl-2-chloro-5-iodonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-5-iodonicotinate, Methyl 2-chloro-5-iodopyridine-3-carboxylate

A104843-100MG A104843-1G A104843-250MG A104843-5G
76796-436EA 5 USD
76796-436 76796-438 76796-440 77666-532
Methyl-2-chloro-5-iodonicotinate 97%
Methyl-2-chloro-5-iodonicotinate

Precaución: for Research Use Only

Formula: C₇H₅ClINO₂
MW: 297.48
MDL Number: MFCD06659607
CAS Number: 78686-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR