Ethyl-4-chlorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-chlorobenzoate, 4-Chlorobenzoic acid ethyl ester

A104629-10G A104629-25G A104629-500G A104629-5G A104629-100G
76796-428EA 6 USD
76796-428 76796-430 76796-432 76796-434 77878-806
Ethyl-4-chlorobenzoate 97%
Ethyl-4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD00013645
CAS Number: 7335-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR