2-Phenoxyethyl isobutyrate 98%

Supplier: Ambeed
Warning

A104169-1000G A104169-100G A104169-500G A104169-25G
76796-406EA 37 USD
76796-406 76796-408 76796-410 77666-504
2-Phenoxyethyl isobutyrate 98%
2-Phenoxyethyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.2536 g/mol
MDL Number: MFCD00027363
CAS Number: 103-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR