Doramapimod 99%

Supplier: Ambeed
Warning

A103377-10MG A103377-1MG A103377-2MG A103377-50MG A103377-5MG A103377-250MG A103377-100MG A103377-1G
76796-356EA 35 USD
76796-356 76796-358 76796-360 76796-362 76796-364 77599-504 77623-854 77623-856
Doramapimod 99%
Doramapimod

Precaución: for Research Use Only

Formula: C₃₁H₃₇N₅O₃
MDL Number: MFCD09752957
CAS Number: 285983-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR