DL-Tartaric acid 97%

Supplier: Ambeed
Warning

A103267-100G A103267-25G A103267-500G A103267-1000G
76796-350EA 6.37 USD
76796-350 76796-352 76796-354 77592-096
DL-Tartaric acid 97%
DL-Tartaric acid

Precaución: for Research Use Only

Formula: C₄H₆O₆
MW: 150.09 g/mol
MDL Number: MFCD00151254
CAS Number: 133-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR