Methyl-2-bromo-5-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-bromo-5-fluorobenzoate
A103056-100G
A103056-10G
A103056-1G
A103056-25G
A103056-500G
A103056-5G
76796-338EA
136.86
USD
76796-338
76796-340
76796-342
76796-344
76796-346
76796-348
Methyl-2-bromo-5-fluorobenzoate 98%
Methyl-2-bromo-5-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ MW: 233.04 g/mol |
MDL Number:
MFCD00086167 CAS Number: 6942-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...