Sodium (2,5-dioxo-4,4-diphenylimidazolidin-1-yl)methyl phosphate 98%

Supplier: Ambeed
Danger

Synonyms: Fosphenytoin disodium

A102515-10MG A102515-1MG A102515-25MG A102515-50MG A102515-5MG
76796-312EA 30.77 USD
76796-312 76796-314 76796-316 76796-318 76796-320
Sodium (2,5-dioxo-4,4-diphenylimidazolidin-1-yl)methyl phosphate 98%
Sodium (2,5-dioxo-4,4-diphenylimidazolidin-1-yl)methyl phosphate

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₂Na₂O₆P
MW: 406.24 g/mol
MDL Number: MFCD00948765
CAS Number: 92134-98-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR