(-)-Dimethyl-D-tartrate 98%

Supplier: Ambeed
Warning

Synonyms: D(-)-Tartaric acid dimethyl ester, (2S,3S)-Dimethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Tartaric acid dimethyl ester, (S,S)-(-)-Dimethyl tartrate, (-)-Dimethyl D-tartrate

A101689-100G A101689-10G A101689-25G A101689-5G
76796-278EA 75 USD
76796-278 76796-280 76796-282 76796-284
(-)-Dimethyl-D-tartrate 98%
(-)-Dimethyl-D-tartrate

Precaución: for Research Use Only

Formula: C₆H₁₀O₆
MDL Number: MFCD00008445
CAS Number: 13171-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR