3-Chloro-5-methylbenzonitrile 97%

Supplier: Ambeed
Danger

A993187-250MG A993187-5G
76792-328EA 27.59 USD
76792-328 76792-330
3-Chloro-5-methylbenzonitrile 97%
3-Chloro-5-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClN
MW: 151.59 g/mol
MDL Number: MFCD09800704
CAS Number: 189161-09-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR