Tris[2-(2-methoxyethoxy)ethyl]amine 95%

Supplier: Ambeed
Warning

Synonyms: Tris(2-(2-methoxyethoxy)ethyl)amine, Tris(3,6-dioxaheptyl)amine, Tris(dioxa-3,6-heptyl)amine

A980432-100G A980432-25G A980432-500G A980432-5G
76792-302EA 95.49 USD
76792-302 76792-304 76792-306 76792-308
Tris[2-(2-methoxyethoxy)ethyl]amine 95%
Tris[2-(2-methoxyethoxy)ethyl]amine

Precaución: for Research Use Only

Formula: C₁₅H₃₃NO₆
MW: 323.43 g/mol
MDL Number: MFCD00010748
CAS Number: 70384-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR