4-Glycidyloxycarbazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2,3-Epoxypropoxy)carbazole, Carvedilol Related Compound D

A978865-25G A978865-5G A978865-100G A978865-500G
76792-298EA 26.53 USD
76792-298 76792-300 77670-426 77670-428
4-Glycidyloxycarbazole 98%
4-Glycidyloxycarbazole

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
MDL Number: MFCD03411884
CAS Number: 51997-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR