tert-Butyl (4-vinylphenyl) carbonate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A977981-1G A977981-25G A977981-5G
76792-286EA 7 USD
76792-286 76792-288 76792-290
tert-Butyl (4-vinylphenyl) carbonate 98% stabilized
tert-Butyl (4-vinylphenyl) carbonate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₃
MDL Number: MFCD00145182
CAS Number: 87188-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR