4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methyl-1-piperazinyl)phenylboronic Acid Pinacol Ester

A975580-100G A975580-100MG A975580-1G A975580-25G A975580-5G A975580-250MG
76792-268EA 715 USD
76792-268 76792-270 76792-272 76792-274 76792-276 76792-278 77873-396
4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester 96%
4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₇BN₂O₂
MW: 302.2195 g/mol
MDL Number: MFCD06795650
CAS Number: 747413-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR