2,5-Dichloro-3-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichloronicotinaldehyde, 2,5-Dichloro-3-formylpyridine

A972773-1G A972773-250MG A972773-5G
76792-262EA 41.38 USD
76792-262 76792-264 76792-266
2,5-Dichloro-3-formylpyridine 98%
2,5-Dichloro-3-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176.0000 g/mol
MDL Number: MFCD08056627
CAS Number: 176433-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR