Tris(trimethylsilyl)phosphate 97%

Supplier: Ambeed
Danger

Synonyms: Tris(trimethylsilyl) phosphate, Phosphoric Acid Tris(trimethylsilyl) Ester

A962485-10G A962485-25G
76792-248EA 17 USD
76792-248 76792-250
Tris(trimethylsilyl)phosphate 97%
Tris(trimethylsilyl)phosphate

Precaución: for Research Use Only

Formula: C₉H₂₇O₄PSi₃
MW: 314.54 g/mol
MDL Number: MFCD00008267
CAS Number: 10497-05-9
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR