Triisopropanolamine 98%

Supplier: Ambeed
Warning

A962422-100G A962422-10G A962422-500G
76792-242EA 7 USD
76792-242 76792-244 76792-246
Triisopropanolamine 98%
Triisopropanolamine

Precaución: for Research Use Only

Formula: C₉H₂₁NO₃
MDL Number: MFCD00004533
CAS Number: 122-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR