Tetrafluoro-p-benzoquinone 98%

Supplier: Ambeed
Warning

Synonyms: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

A960703-1G A960703-250MG A960703-5G
76792-238EA 61.54 USD
76792-238 76792-240 77573-900
Tetrafluoro-p-benzoquinone 98%
Tetrafluoro-p-benzoquinone

Precaución: for Research Use Only

Formula: C₆F₄O₂
MW: 180.06 g/mol
MDL Number: MFCD00001592
CAS Number: 527-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR