Triethylene glycol dimethyl ether 99% stabilized
Supplier: Ambeed
|
Danger
|
Synonyms:
2,5,8,11-Tetraoxadodecane
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A958537-25G
A958537-100G
76792-236EA
5.31
USD
76792-236
77670-404
Triethylene glycol dimethyl ether 99% stabilized
Triethylene glycol dimethyl ether
Precaución: for Research Use Only
Formula:
C₈H₁₈O₄ MW: 178.23 g/mol |
MDL Number:
MFCD00008504 CAS Number: 112-49-2 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...