2-Phenyl-2',4',6'-trihydroxyacetophenone 96%

Supplier: Ambeed
Danger

Synonyms: 2',4',6'-Trihydroxy-2-phenylacetophenone, 2-Phenyl-1-(2,4,6-trihydroxyphenyl)ethanone, Benzyl 2,4,6-Trihydroxyphenyl ketone

A956263-100MG A956263-1G A956263-250MG
76792-228EA 22 USD
76792-228 76792-230 76792-232
2-Phenyl-2',4',6'-trihydroxyacetophenone 96%
2-Phenyl-2',4',6'-trihydroxyacetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
MW: 244.2427 g/mol
MDL Number: MFCD00205539
CAS Number: 727-71-9
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR