tert-Butyl 3-fluoro-4-nitrobenzoate 95%

Supplier: Ambeed
Warning

A954483-100MG A954483-1G A954483-250MG A954483-5G
76792-222EA 7 USD
76792-222 76792-224 76792-226 77670-398
tert-Butyl 3-fluoro-4-nitrobenzoate 95%
tert-Butyl 3-fluoro-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₄
MDL Number: MFCD08703397
CAS Number: 157665-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR