N-(2,6-Diethylphenyl)maleimide 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2,6-Diethylphenyl)-1H-pyrrole-2,5-dione

A952667-100MG A952667-1G A952667-250MG
76792-198EA 20 USD
76792-198 76792-200 76792-202
N-(2,6-Diethylphenyl)maleimide 95%
N-(2,6-Diethylphenyl)maleimide

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₂
MDL Number: MFCD00173826
CAS Number: 38167-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR