tert-Butyl-4-chloro-3-oxobutanoate 95%

Supplier: Ambeed
Danger

A941939-100MG A941939-250MG A941939-5G A941939-1G
76792-174EA 62.6 USD
76792-174 76792-176 77595-526 77670-378
tert-Butyl-4-chloro-3-oxobutanoate 95%
tert-Butyl-4-chloro-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₈H₁₃ClO₃
MW: 192.64 g/mol
MDL Number: MFCD00052188
CAS Number: 74530-56-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR