trans-3-Hexenedioic acid 98%

Supplier: Ambeed
Warning

Synonyms: trans-beta-Hydromuconic Acid, trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, Hex-3-enedioic acid

A938367-25G A938367-5G A938367-1G
76792-158EA 251.44 USD
76792-158 77603-322 77670-372
trans-3-Hexenedioic acid 98%
trans-3-Hexenedioic acid

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00002785
CAS Number: 4436-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR