Tetramethylammonium-p-toluenesulfonate 98%

Supplier: Ambeed
Warning

A935990-100G A935990-5G A935990-25G
76792-154EA 60 USD
76792-154 76792-156 77873-380
Tetramethylammonium-p-toluenesulfonate 98%
Tetramethylammonium-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃S
MDL Number: MFCD00043173
CAS Number: 3983-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR