Trimellitic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,4-Benzenetricarboxylic acid, Benzene-1,2,4-tricarboxylic acid

A927137-100G A927137-25G A927137-500G A927137-5G
76792-142EA 50.93 USD
76792-142 76792-144 76792-146 76792-148
Trimellitic acid 98%
Trimellitic acid

Precaución: for Research Use Only

Formula: C₉H₆O₆
MW: 210.14 g/mol
MDL Number: MFCD00002470
CAS Number: 528-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR