Khellin 95%

Supplier: Ambeed
Danger

Synonyms: Visammin, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one

A925735-100MG A925735-1G A925735-1MG
76792-136EA 24.41 USD
76792-136 76792-138 76792-140
Khellin 95%
Khellin

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₅
MW: 260.25 g/mol
MDL Number: MFCD00005007
CAS Number: 82-02-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR