Tetraphenylphosphonium tetraphenylborate 98%

Supplier: Ambeed
Warning

A925093-1G A925093-25G A925093-5G
76792-130EA 6 USD
76792-130 76792-132 76792-134
Tetraphenylphosphonium tetraphenylborate 98%
Tetraphenylphosphonium tetraphenylborate

Precaución: for Research Use Only

Formula: C₄₈H₄₀BP
MW: 658.6160 g/mol
MDL Number: MFCD00145389
CAS Number: 15525-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR