Tetramethylammonium hexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: Tetramethylammoniumhexafluorophosphate

A919805-100G A919805-25G A919805-5G
76792-124EA 55 USD
76792-124 76792-126 76792-128
Tetramethylammonium hexafluorophosphate 98%
Tetramethylammonium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₄H₁₂F₆NP
MDL Number: MFCD00012012
CAS Number: 558-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR