N,N'-Ethylenedianiline 98%

Supplier: Ambeed
Warning

Synonyms: N1,N2-Diphenylethane-1,2-diamine, 1,2-Dianilinoethane, N,N'-Diphenylethylenediamine

A915482-25G A915482-5G A915482-1G A915482-100G
76792-120EA 78 USD
76792-120 76792-122 77603-290 77873-368
N,N'-Ethylenedianiline 98%
N,N'-Ethylenedianiline

Precaución: for Research Use Only

Formula: C₆H₅NHCH₂CH₂NHC₆H₅
MDL Number: MFCD00003019
CAS Number: 150-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR