Tiliroside 95%

Supplier: Ambeed
Warning

Synonyms: ((2R,3S,4S,5R,6S)-6-((5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-3-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)methyl (E)-3-(4-hydroxyphenyl)acrylate

A913539-10MG A913539-1MG A913539-25MG A913539-50MG A913539-5MG
76792-110EA 78.51 USD
76792-110 76792-112 76792-114 76792-116 76792-118
Tiliroside 95%
Tiliroside

Precaución: for Research Use Only

Formula: C₃₀H₂₆O₁₃
MW: 594.525 g/mol
MDL Number: MFCD00017454
CAS Number: 20316-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR