Methyl 3,5-bis(benzyloxy)benzoate 97%

Supplier: Ambeed
Warning

A909288-1G A909288-25G A909288-5G
76792-098EA 20.16 USD
76792-098 76792-100 76792-102
Methyl 3,5-bis(benzyloxy)benzoate 97%
Methyl 3,5-bis(benzyloxy)benzoate

Precaución: for Research Use Only

Formula: C₂₂H₂₀O₄
MW: 348.39 g/mol
MDL Number: MFCD02728721
CAS Number: 58605-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR