6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A906988-100MG A906988-250MG
76792-096EA 135 USD
76792-096 77873-360
6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzo[d]imidazole 97%
6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₄H₉Cl₃N₂O₂S
MDL Number: MFCD19441813
CAS Number: 100648-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR