Triethylene glycol monobutyl ether 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Triethyleneglycolmonobutylether, Butyl Triglycol

A906941-1000G A906941-100G A906941-500G
76792-090EA 23.34 USD
76792-090 76792-092 76792-094
Triethylene glycol monobutyl ether 98% (by GC)
Triethylene glycol monobutyl ether

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₄
MW: 206.28 g/mol
MDL Number: MFCD00020606
CAS Number: 143-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR