tert-Butyl acetoacetate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl Acetoacetate
A902589-100G
A902589-25G
A902589-500G
A902589-1000G
76792-084EA
15
USD
76792-084
76792-086
76792-088
77552-698
tert-Butyl acetoacetate 97%
tert-Butyl acetoacetate
Precaución: for Research Use Only
Formula:
C₈H₁₄O₃ |
MDL Number:
MFCD00008811 CAS Number: 1694-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...